Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 138090 | ||||||||||||||||||||
Literature Links: |
H6PD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
H6PD - hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282587.1 | 41 | Intron | NP_001269516.1 | |||
NM_004285.3 | 41 | UTR 5 | NP_004276.2 | |||
XM_005263540.4 | 41 | Intron | XP_005263597.1 | |||
XM_006711052.3 | 41 | UTR 5 | XP_006711115.1 | |||
XM_017002865.1 | 41 | Intron | XP_016858354.1 | |||
XM_017002866.1 | 41 | Intron | XP_016858355.1 |
Set Membership: |
JSNP |