Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191350 MIM: 601772 MIM: 609806 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPAGT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DPAGT1 - dolichyl-phosphate N-acetylglucosaminephosphotransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
H2AFX - H2A histone family member X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002105.2 | 1214 | UTR 3 | NP_002096.1 |
HMBS - hydroxymethylbilane synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000190.3 | 1214 | Intron | NP_000181.2 | |||
NM_001024382.1 | 1214 | Intron | NP_001019553.1 | |||
NM_001258208.1 | 1214 | Intron | NP_001245137.1 | |||
NM_001258209.1 | 1214 | Intron | NP_001245138.1 | |||
XM_005271531.1 | 1214 | Intron | XP_005271588.1 | |||
XM_005271532.1 | 1214 | Intron | XP_005271589.1 | |||
XM_005271533.3 | 1214 | Intron | XP_005271590.1 | |||
XM_011542796.1 | 1214 | Intron | XP_011541098.1 | |||
XM_017017629.1 | 1214 | Intron | XP_016873118.1 |