Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR160 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPR160 - G protein-coupled receptor 160 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014373.2 | 92 | Intron | NP_055188.1 | |||
XM_005247346.3 | 92 | Intron | XP_005247403.1 | |||
XM_005247347.3 | 92 | Intron | XP_005247404.1 | |||
XM_006713582.3 | 92 | UTR 5 | XP_006713645.1 | |||
XM_011512678.2 | 92 | UTR 5 | XP_011510980.1 | |||
XM_017006161.1 | 92 | Intron | XP_016861650.1 | |||
XM_017006162.1 | 92 | Intron | XP_016861651.1 | |||
XM_017006163.1 | 92 | Intron | XP_016861652.1 |
Set Membership: |
HapMap |