Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611230 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMF2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LMF2 - lipase maturation factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033200.2 | 54 | Intron | NP_149977.2 | |||
XM_005261955.3 | 54 | Intron | XP_005262012.1 | |||
XM_006724426.3 | 54 | Intron | XP_006724489.1 | |||
XM_006724427.3 | 54 | Intron | XP_006724490.1 | |||
XM_017029077.1 | 54 | Intron | XP_016884566.1 |
NCAPH2 - non-SMC condensin II complex subunit H2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185011.1 | 54 | UTR 5 | NP_001171940.1 | |||
NM_014551.4 | 54 | UTR 5 | NP_055366.3 | |||
NM_152299.3 | 54 | UTR 5 | NP_689512.2 | |||
XM_005261912.4 | 54 | UTR 5 | XP_005261969.1 | |||
XM_011530685.2 | 54 | Intron | XP_011528987.1 | |||
XM_017028793.1 | 54 | UTR 5 | XP_016884282.1 |