Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606607 MIM: 606472 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMA7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMA7 - proteasome subunit alpha 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SS18L1 - SS18L1, nBAF chromatin remodeling complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301778.1 | Intron | NP_001288707.1 | ||||
NM_198935.2 | Intron | NP_945173.1 | ||||
XM_005260389.3 | Intron | XP_005260446.1 | ||||
XM_005260391.1 | Intron | XP_005260448.1 | ||||
XM_011528764.2 | Intron | XP_011527066.1 | ||||
XM_011528767.1 | Intron | XP_011527069.1 | ||||
XM_017027784.1 | Intron | XP_016883273.1 |