Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGTCATTGCTGGAACCAGCGGC[C/T]TCCATGTGAGGTACAGGGGAACGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103180 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARF1 - ADP ribosylation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024226.1 | Intron | NP_001019397.1 | ||||
NM_001024227.1 | Intron | NP_001019398.1 | ||||
NM_001024228.1 | Intron | NP_001019399.1 | ||||
NM_001658.3 | Intron | NP_001649.1 | ||||
XM_017001233.1 | Intron | XP_016856722.1 |
LOC100506571 - uncharacterized LOC100506571 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3620 - microRNA 3620 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP |