Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601874 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELL2 - elongation factor for RNA polymerase II 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012081.5 | Intron | NP_036213.2 | ||||
XM_006714575.2 | Intron | XP_006714638.1 | ||||
XM_017009239.1 | Intron | XP_016864728.1 | ||||
XM_017009240.1 | Intron | XP_016864729.1 | ||||
XM_017009241.1 | Intron | XP_016864730.1 | ||||
XM_017009242.1 | Intron | XP_016864731.1 | ||||
XM_017009243.1 | Intron | XP_016864732.1 |