Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605009 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADAMTS7 - ADAM metallopeptidase with thrombospondin type 1 motif 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014272.4 | 2376 | Silent Mutation | GGA,GGG | G,G 711 | NP_055087.2 | |
XM_005254137.3 | 2376 | Silent Mutation | GGA,GGG | G,G 711 | XP_005254194.1 | |
XM_011521166.2 | 2376 | Intron | XP_011519468.1 |
Set Membership: |
HapMap |