Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159556 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLLT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MLLT1 - MLLT1, super elongation complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005934.3 | Intron | NP_005925.2 | ||||
XM_011528021.1 | Intron | XP_011526323.1 | ||||
XM_011528022.1 | Intron | XP_011526324.1 | ||||
XM_011528023.1 | Intron | XP_011526325.1 | ||||
XM_017026819.1 | Intron | XP_016882308.1 |
Set Membership: |
HapMap |