Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608377 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFGAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARFGAP1 - ADP ribosylation factor GTPase activating protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281482.1 | Intron | NP_001268411.1 | ||||
NM_001281483.1 | Intron | NP_001268412.1 | ||||
NM_001281484.1 | Intron | NP_001268413.1 | ||||
NM_018209.3 | Intron | NP_060679.1 | ||||
NM_175609.2 | Intron | NP_783202.1 | ||||
XM_006723823.2 | Intron | XP_006723886.1 | ||||
XM_011528901.1 | Intron | XP_011527203.1 | ||||
XM_017027950.1 | Intron | XP_016883439.1 |