Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGACCCTTGCCTTTTTGTTTTC[C/T]TCTGTTACAATGTGCAGTAAGTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609361 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPE1-MOB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HSPE1-MOB4 - HSPE1-MOB4 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202485.1 | Intron | NP_001189414.1 |
MOB4 - MOB family member 4, phocein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100819.2 | Intron | NP_001094289.1 | ||||
NM_001204094.1 | Intron | NP_001191023.1 | ||||
NM_015387.4 | Intron | NP_056202.2 | ||||
NM_199482.3 | Intron | NP_955776.1 |
Set Membership: |
HapMap |