Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613114 MIM: 608694 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM134B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM134B - family with sequence similarity 134 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034850.2 | 2397 | UTR 3 | NP_001030022.1 | |||
NM_019000.4 | 2397 | UTR 3 | NP_061873.2 | |||
XM_011514053.2 | 2397 | UTR 3 | XP_011512355.1 | |||
XM_011514054.2 | 2397 | UTR 3 | XP_011512356.1 | |||
XM_011514055.2 | 2397 | UTR 3 | XP_011512357.1 |
ZNF622 - zinc finger protein 622 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |