Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTTCAACTGCATGCTAACACAG[A/G]CTGCTGTGAGGGAAACTGAACAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611587 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP19 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGAP19 - Rho GTPase activating protein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204300.1 | 4574 | UTR 3 | NP_001191229.1 | |||
NM_001256423.1 | 4574 | UTR 3 | NP_001243352.1 | |||
NM_032900.5 | 4574 | UTR 3 | NP_116289.4 |
ARHGAP19-SLIT1 - ARHGAP19-SLIT1 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |