Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAGCCTTCCACCACCCAACCCA[A/G]GTCCTGGACCTTGTCATTAGGGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611121 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLMN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLMN - calmin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024734.3 | 9520 | UTR 3 | NP_079010.2 | |||
XM_011537158.1 | 9520 | UTR 3 | XP_011535460.1 | |||
XM_011537159.2 | 9520 | Intron | XP_011535461.1 | |||
XM_017021646.1 | 9520 | UTR 3 | XP_016877135.1 | |||
XM_017021647.1 | 9520 | Intron | XP_016877136.1 |
DICER1-AS1 - DICER1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |