Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607198 | ||||||||||||||||||||
Literature Links: |
EFCAB11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EFCAB11 - EF-hand calcium binding domain 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284266.1 | 706 | Intron | NP_001271195.1 | |||
NM_001284267.1 | 706 | Intron | NP_001271196.1 | |||
NM_001284268.1 | 706 | Intron | NP_001271197.1 | |||
NM_001284269.1 | 706 | Intron | NP_001271198.1 | |||
NM_145231.3 | 706 | Intron | NP_660274.1 |
TDP1 - tyrosyl-DNA phosphodiesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008744.1 | 706 | Intron | NP_001008744.1 | |||
NM_018319.3 | 706 | Intron | NP_060789.2 | |||
XM_005267847.2 | 706 | UTR 5 | XP_005267904.1 | |||
XM_005267848.2 | 706 | Intron | XP_005267905.1 | |||
XM_006720197.3 | 706 | Intron | XP_006720260.1 | |||
XM_006720198.3 | 706 | Intron | XP_006720261.1 | |||
XM_006720199.1 | 706 | Intron | XP_006720262.1 | |||
XM_011536942.2 | 706 | UTR 5 | XP_011535244.1 | |||
XM_011536943.2 | 706 | Intron | XP_011535245.1 | |||
XM_011536944.1 | 706 | Intron | XP_011535246.1 | |||
XM_017021439.1 | 706 | Intron | XP_016876928.1 | |||
XM_017021440.1 | 706 | Intron | XP_016876929.1 | |||
XM_017021441.1 | 706 | Intron | XP_016876930.1 | |||
XM_017021442.1 | 706 | Intron | XP_016876931.1 | |||
XM_017021443.1 | 706 | Intron | XP_016876932.1 | |||
XM_017021444.1 | 706 | Intron | XP_016876933.1 | |||
XM_017021445.1 | 706 | Intron | XP_016876934.1 | |||
XM_017021446.1 | 706 | Intron | XP_016876935.1 |