Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605908 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MLC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MLC1 - megalencephalic leukoencephalopathy with subcortical cysts 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015166.3 | Intron | NP_055981.1 | ||||
NM_139202.2 | Intron | NP_631941.1 | ||||
XM_011530678.1 | Intron | XP_011528980.1 | ||||
XM_017028671.1 | Intron | XP_016884160.1 |
TTLL8 - tubulin tyrosine ligase like 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |