Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCTGGGCTCTCACCTGCACAAA[A/G]GCATATGCTAGGTTAGAGGGGTAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 102645 MIM: 142408 MIM: 614472 | ||||||||||||||||||||
Literature Links: |
APEH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APEH - acylaminoacyl-peptide hydrolase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MST1 - macrophage stimulating 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020998.3 | Intron | NP_066278.3 | ||||
XM_006713166.1 | Intron | XP_006713229.1 | ||||
XM_011533732.1 | Intron | XP_011532034.1 | ||||
XM_011533737.2 | Intron | XP_011532039.1 | ||||
XM_011533738.2 | Intron | XP_011532040.1 | ||||
XM_017006460.1 | Intron | XP_016861949.1 | ||||
XM_017006461.1 | Intron | XP_016861950.1 | ||||
XM_017006462.1 | Intron | XP_016861951.1 | ||||
XM_017006463.1 | Intron | XP_016861952.1 | ||||
XM_017006464.1 | Intron | XP_016861953.1 | ||||
XM_017006465.1 | Intron | XP_016861954.1 | ||||
XM_017006466.1 | Intron | XP_016861955.1 | ||||
XM_017006467.1 | Intron | XP_016861956.1 | ||||
XM_017006468.1 | Intron | XP_016861957.1 |
RNF123 - ring finger protein 123 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022064.4 | Intron | NP_071347.2 | ||||
XM_006713285.1 | Intron | XP_006713348.1 | ||||
XM_011533995.1 | Intron | XP_011532297.1 | ||||
XM_017007018.1 | Intron | XP_016862507.1 |