Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 610389 MIM: 612942 MIM: 605440 | ||||||||||||||||||||||||||
Literature Links: |
LAMTOR2 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
LAMTOR2 - late endosomal/lysosomal adaptor, MAPK and MTOR activator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145264.1 | Intron | NP_001138736.1 | ||||
NM_014017.3 | Intron | NP_054736.1 |
RAB25 - RAB25, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBQLN4 - ubiquilin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304342.1 | Intron | NP_001291271.1 | ||||
NM_020131.4 | Intron | NP_064516.2 | ||||
XM_005245348.3 | Intron | XP_005245405.1 |
Set Membership: |
HapMap JSNP |