Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600493 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRE1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADGRE1 - adhesion G protein-coupled receptor E1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256252.1 | Intron | NP_001243181.1 | ||||
NM_001256253.1 | Intron | NP_001243182.1 | ||||
NM_001256254.1 | Intron | NP_001243183.1 | ||||
NM_001256255.1 | Intron | NP_001243184.1 | ||||
NM_001974.4 | Intron | NP_001965.3 | ||||
XM_011527794.1 | Intron | XP_011526096.1 |
Set Membership: |
HapMap |