Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300970 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MORC4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MORC4 - MORC family CW-type zinc finger 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085354.2 | 2232 | Missense Mutation | CGC,TGC | R,C 653 | NP_001078823.1 | |
NM_024657.4 | 2232 | Missense Mutation | CGC,TGC | R,C 653 | NP_078933.3 | |
XM_005262190.3 | 2232 | Missense Mutation | CGC,TGC | R,C 561 | XP_005262247.1 | |
XM_006724691.2 | 2232 | Missense Mutation | CGC,TGC | R,C 531 | XP_006724754.1 | |
XM_011531027.2 | 2232 | Missense Mutation | CGC,TGC | R,C 476 | XP_011529329.1 | |
XM_017029844.1 | 2232 | Missense Mutation | CGC,TGC | R,C 561 | XP_016885333.1 |