Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605604 MIM: 603790 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAIP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PAIP2 - poly(A) binding protein interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033112.2 | 1955 | Silent Mutation | TAC,TAT | Y,Y 124 | NP_001028284.1 | |
NM_016480.4 | 1955 | Silent Mutation | TAC,TAT | Y,Y 124 | NP_057564.3 | |
XM_011543428.1 | 1955 | Silent Mutation | TAC,TAT | Y,Y 124 | XP_011541730.1 | |
XM_017009522.1 | 1955 | Silent Mutation | TAC,TAT | Y,Y 124 | XP_016865011.1 | |
XM_017009523.1 | 1955 | Silent Mutation | TAC,TAT | Y,Y 124 | XP_016865012.1 |
SLC23A1 - solute carrier family 23 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005847.4 | 1955 | Intron | NP_005838.3 | |||
NM_152685.3 | 1955 | Intron | NP_689898.2 | |||
XM_005272148.3 | 1955 | Intron | XP_005272205.3 | |||
XM_005272149.3 | 1955 | Intron | XP_005272206.3 | |||
XM_006714741.2 | 1955 | Intron | XP_006714804.2 | |||
XM_011543765.2 | 1955 | Intron | XP_011542067.1 | |||
XM_011543766.1 | 1955 | Intron | XP_011542068.1 | |||
XM_011543767.1 | 1955 | Intron | XP_011542069.1 | |||
XM_011543768.1 | 1955 | Intron | XP_011542070.1 | |||
XM_011543769.1 | 1955 | Intron | XP_011542071.1 |
Set Membership: |
HapMap |