Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602341 MIM: 614085 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FOXM1 - forkhead box M1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243088.1 | Intron | NP_001230017.1 | ||||
NM_001243089.1 | Intron | NP_001230018.1 | ||||
NM_021953.3 | Intron | NP_068772.2 | ||||
NM_202002.2 | Intron | NP_973731.1 | ||||
NM_202003.2 | Intron | NP_973732.1 | ||||
XM_005253676.3 | Intron | XP_005253733.1 | ||||
XM_011520930.2 | Intron | XP_011519232.1 | ||||
XM_011520931.2 | Intron | XP_011519233.1 | ||||
XM_011520932.1 | Intron | XP_011519234.1 | ||||
XM_011520933.1 | Intron | XP_011519235.1 | ||||
XM_011520934.2 | Intron | XP_011519236.1 | ||||
XM_011520935.1 | Intron | XP_011519237.1 |
RHNO1 - RAD9-HUS1-RAD1 interacting nuclear orphan 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252499.2 | Intron | NP_001239428.1 | ||||
NM_001252500.2 | Intron | NP_001239429.1 | ||||
NM_001257097.1 | Intron | NP_001244026.1 | ||||
NM_001257098.1 | Intron | NP_001244027.1 |