Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126451 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DRD3 - dopamine receptor D3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000796.5 | 1734 | Intron | NP_000787.2 | |||
NM_001282563.2 | 1734 | Intron | NP_001269492.1 | |||
NM_001290809.1 | 1734 | Intron | NP_001277738.1 | |||
NM_033663.5 | 1734 | Intron | NP_387512.3 | |||
XM_017005829.1 | 1734 | UTR 3 | XP_016861318.1 |
Set Membership: |
HapMap |