Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608064 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLHL5 - kelch like family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007075.2 | Intron | NP_001007076.1 | ||||
NM_001171654.1 | Intron | NP_001165125.1 | ||||
NM_015990.4 | Intron | NP_057074.3 | ||||
NM_199039.3 | Intron | NP_950240.2 | ||||
XM_005262655.4 | Intron | XP_005262712.1 | ||||
XM_011513699.2 | Intron | XP_011512001.1 | ||||
XM_011513700.2 | Intron | XP_011512002.1 | ||||
XM_011513701.2 | Intron | XP_011512003.1 | ||||
XM_017008273.1 | Intron | XP_016863762.1 | ||||
XM_017008274.1 | Intron | XP_016863763.1 | ||||
XM_017008275.1 | Intron | XP_016863764.1 | ||||
XM_017008276.1 | Intron | XP_016863765.1 |
Set Membership: |
HapMap |