Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611212 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf19 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C4orf19 - chromosome 4 open reading frame 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104629.1 | 1068 | Missense Mutation | GAG,GGG | E,G 274 | NP_001098099.1 | |
NM_018302.2 | 1068 | Missense Mutation | GAG,GGG | E,G 274 | NP_060772.2 | |
XM_011513712.2 | 1068 | Missense Mutation | GAG,GGG | E,G 274 | XP_011512014.1 | |
XM_011513713.2 | 1068 | Missense Mutation | GAG,GGG | E,G 274 | XP_011512015.1 | |
XM_017008361.1 | 1068 | Missense Mutation | GAG,GGG | E,G 274 | XP_016863850.1 |
RELL1 - RELT like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085399.1 | 1068 | UTR 3 | NP_001078868.1 | |||
NM_001085400.1 | 1068 | Intron | NP_001078869.1 | |||
XM_017008590.1 | 1068 | Intron | XP_016864079.1 |