Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604322 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC17A5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC17A5 - solute carrier family 17 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012434.4 | 1619 | UTR 3 | NP_036566.1 | |||
XM_005248710.3 | 1619 | UTR 3 | XP_005248767.1 | |||
XM_005248711.2 | 1619 | UTR 3 | XP_005248768.1 | |||
XM_011535750.2 | 1619 | UTR 3 | XP_011534052.1 | |||
XM_017010730.1 | 1619 | UTR 3 | XP_016866219.1 | |||
XM_017010731.1 | 1619 | UTR 3 | XP_016866220.1 |
Set Membership: |
HapMap |