Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609752 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCOA7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NCOA7 - nuclear receptor coactivator 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122842.2 | 180 | Intron | NP_001116314.1 | |||
NM_001199619.1 | 180 | UTR 5 | NP_001186548.1 | |||
NM_001199620.1 | 180 | UTR 5 | NP_001186549.1 | |||
NM_001199621.1 | 180 | Intron | NP_001186550.1 | |||
NM_001199622.1 | 180 | Intron | NP_001186551.1 | |||
NM_181782.4 | 180 | Intron | NP_861447.3 | |||
XM_005266822.4 | 180 | Intron | XP_005266879.1 | |||
XM_006715340.3 | 180 | UTR 5 | XP_006715403.1 | |||
XM_011535455.2 | 180 | Intron | XP_011533757.1 | |||
XM_017010269.1 | 180 | Intron | XP_016865758.1 | |||
XM_017010270.1 | 180 | UTR 5 | XP_016865759.1 | |||
XM_017010271.1 | 180 | Intron | XP_016865760.1 | |||
XM_017010272.1 | 180 | Intron | XP_016865761.1 | |||
XM_017010273.1 | 180 | Intron | XP_016865762.1 | |||
XM_017010274.1 | 180 | UTR 5 | XP_016865763.1 |