Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611864 MIM: 612334 | ||||||||||||||||||||
Literature Links: |
ARMC10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARMC10 - armadillo repeat containing 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAPEPLD - N-acyl phosphatidylethanolamine phospholipase D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122838.1 | 4055 | UTR 3 | NP_001116310.1 | |||
NM_198990.4 | 4055 | UTR 3 | NP_945341.3 | |||
XM_005250214.4 | 4055 | UTR 3 | XP_005250271.2 | |||
XM_005250215.2 | 4055 | UTR 3 | XP_005250272.1 | |||
XM_005250218.2 | 4055 | UTR 3 | XP_005250275.1 | |||
XM_011515933.2 | 4055 | UTR 3 | XP_011514235.1 | |||
XM_011515934.2 | 4055 | UTR 3 | XP_011514236.1 | |||
XM_017011854.1 | 4055 | UTR 3 | XP_016867343.1 | |||
XM_017011855.1 | 4055 | UTR 3 | XP_016867344.1 | |||
XM_017011856.1 | 4055 | Intron | XP_016867345.1 | |||
XM_017011857.1 | 4055 | Intron | XP_016867346.1 |