Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602105 MIM: 179080 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSH4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MSH4 - mutS homolog 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RABGGTB - Rab geranylgeranyltransferase beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004582.3 | 129 | Silent Mutation | ACA,ACT | T,T 17 | NP_004573.2 |
SNORD45A - small nucleolar RNA, C/D box 45A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD45B - small nucleolar RNA, C/D box 45B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD45C - small nucleolar RNA, C/D box 45C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP |