Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616064 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TINAGL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TINAGL1 - tubulointerstitial nephritis antigen like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204414.1 | Intron | NP_001191343.1 | ||||
NM_001204415.1 | Intron | NP_001191344.1 | ||||
NM_022164.2 | Intron | NP_071447.1 | ||||
XM_005271106.2 | Intron | XP_005271163.1 | ||||
XM_005271107.2 | Intron | XP_005271164.1 | ||||
XM_011541946.1 | Intron | XP_011540248.1 |
Set Membership: |
HapMap JSNP |