Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614126 MIM: 614127 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MARC1 - mitochondrial amidoxime reducing component 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022746.3 | 202 | UTR 5 | NP_073583.3 | |||
XM_011509900.2 | 202 | UTR 5 | XP_011508202.1 | |||
XM_011509903.2 | 202 | Intron | XP_011508205.1 | |||
XM_011509904.2 | 202 | Intron | XP_011508206.1 | |||
XM_017002096.1 | 202 | Intron | XP_016857585.1 | |||
XM_017002097.1 | 202 | Intron | XP_016857586.1 |
MARC2 - mitochondrial amidoxime reducing component 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |