Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609749 | ||||||||||||||||||||
Literature Links: |
UXS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UXS1 - UDP-glucuronate decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253875.1 | 1663 | UTR 3 | NP_001240804.1 | |||
NM_001253876.1 | 1663 | UTR 3 | NP_001240805.1 | |||
NM_025076.4 | 1663 | UTR 3 | NP_079352.2 | |||
XM_006712775.3 | 1663 | UTR 3 | XP_006712838.1 | |||
XM_011511902.2 | 1663 | UTR 3 | XP_011510204.2 | |||
XM_011511903.2 | 1663 | UTR 3 | XP_011510205.2 | |||
XM_011511904.2 | 1663 | UTR 3 | XP_011510206.1 | |||
XM_017005008.1 | 1663 | UTR 3 | XP_016860497.1 | |||
XM_017005009.1 | 1663 | UTR 3 | XP_016860498.1 | |||
XM_017005010.1 | 1663 | UTR 3 | XP_016860499.1 | |||
XM_017005011.1 | 1663 | UTR 3 | XP_016860500.1 | |||
XM_017005012.1 | 1663 | UTR 3 | XP_016860501.1 | |||
XM_017005013.1 | 1663 | UTR 3 | XP_016860502.1 | |||
XM_017005014.1 | 1663 | UTR 3 | XP_016860503.1 |