Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604661 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf76 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C10orf76 - chromosome 10 open reading frame 76 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024541.2 | Intron | NP_078817.2 | ||||
XM_006717969.1 | Intron | XP_006718032.1 | ||||
XM_011540151.2 | Intron | XP_011538453.1 | ||||
XM_011540152.2 | Intron | XP_011538454.1 | ||||
XM_011540153.2 | Intron | XP_011538455.1 | ||||
XM_011540154.2 | Intron | XP_011538456.1 | ||||
XM_011540156.2 | Intron | XP_011538458.1 | ||||
XM_011540157.2 | Intron | XP_011538459.1 | ||||
XM_017016635.1 | Intron | XP_016872124.1 | ||||
XM_017016636.1 | Intron | XP_016872125.1 |
KCNIP2 - potassium voltage-gated channel interacting protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |