Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605696 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNG2 - potassium voltage-gated channel modifier subfamily G member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012283.1 | 1872 | Intron | NP_036415.1 |
PQLC1 - PQ loop repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146343.1 | 1872 | UTR 3 | NP_001139815.1 | |||
NM_001146345.1 | 1872 | UTR 3 | NP_001139817.1 | |||
NM_025078.4 | 1872 | UTR 3 | NP_079354.2 | |||
XM_005266770.2 | 1872 | UTR 3 | XP_005266827.1 | |||
XM_011526188.1 | 1872 | UTR 3 | XP_011524490.1 | |||
XM_017025997.1 | 1872 | UTR 3 | XP_016881486.1 | |||
XM_017025998.1 | 1872 | UTR 3 | XP_016881487.1 | |||
XM_017025999.1 | 1872 | Intron | XP_016881488.1 | |||
XM_017026000.1 | 1872 | Intron | XP_016881489.1 | |||
XM_017026001.1 | 1872 | UTR 3 | XP_016881490.1 | |||
XM_017026002.1 | 1872 | Intron | XP_016881491.1 | |||
XM_017026003.1 | 1872 | UTR 3 | XP_016881492.1 | |||
XM_017026004.1 | 1872 | UTR 3 | XP_016881493.1 | |||
XM_017026005.1 | 1872 | UTR 3 | XP_016881494.1 |
Set Membership: |
HapMap |