Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611389 MIM: 616168 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRICKLE4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRICKLE4 - prickle planar cell polarity protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOMM6 - translocase of outer mitochondrial membrane 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP49 - ubiquitin specific peptidase 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286554.1 | 6068 | UTR 3 | NP_001273483.1 | |||
NM_018561.4 | 6068 | Intron | NP_061031.2 |
Set Membership: |
HapMap |