Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614169 MIM: 612778 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NBEAL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NBEAL2 - neurobeachin like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRADDP - neurotrophin receptor associated death domain, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETD2 - SET domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014159.6 | Intron | NP_054878.5 | ||||
XM_011533631.2 | Intron | XP_011531933.1 | ||||
XM_011533632.2 | Intron | XP_011531934.1 | ||||
XM_011533633.2 | Intron | XP_011531935.1 | ||||
XM_011533634.1 | Intron | XP_011531936.1 | ||||
XM_017006270.1 | Intron | XP_016861759.1 |