Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNJL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCNJL - cyclin J like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308173.1 | 2619 | UTR 3 | NP_001295102.1 | |||
NM_024565.6 | 2619 | UTR 3 | NP_078841.3 | |||
XM_006714917.3 | 2619 | UTR 3 | XP_006714980.1 | |||
XM_011534646.2 | 2619 | UTR 3 | XP_011532948.1 | |||
XM_011534647.2 | 2619 | UTR 3 | XP_011532949.1 | |||
XM_017009846.1 | 2619 | UTR 3 | XP_016865335.1 | |||
XM_017009847.1 | 2619 | UTR 3 | XP_016865336.1 | |||
XM_017009848.1 | 2619 | UTR 3 | XP_016865337.1 | |||
XM_017009849.1 | 2619 | UTR 3 | XP_016865338.1 |
Set Membership: |
HapMap |