Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602618 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CTBP1 - C-terminal binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012614.1 | 162 | Intron | NP_001012632.1 | |||
NM_001328.2 | 162 | Intron | NP_001319.1 | |||
XM_005272261.3 | 162 | Intron | XP_005272318.1 | |||
XM_005272263.4 | 162 | Intron | XP_005272320.1 | |||
XM_011513395.2 | 162 | Intron | XP_011511697.1 | |||
XM_011513396.2 | 162 | Intron | XP_011511698.1 | |||
XM_011513397.2 | 162 | Intron | XP_011511699.1 | |||
XM_017007762.1 | 162 | Intron | XP_016863251.1 | |||
XM_017007763.1 | 162 | UTR 5 | XP_016863252.1 | |||
XM_017007764.1 | 162 | Intron | XP_016863253.1 | |||
XM_017007765.1 | 162 | Intron | XP_016863254.1 | |||
XM_017007766.1 | 162 | Intron | XP_016863255.1 | |||
XM_017007767.1 | 162 | Intron | XP_016863256.1 |
CTBP1-AS2 - CTBP1 antisense RNA 2 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |