Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602315 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP2K3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAP2K3 - mitogen-activated protein kinase kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316332.1 | Intron | NP_001303261.1 | ||||
NM_002756.4 | Intron | NP_002747.2 | ||||
NM_145109.2 | Intron | NP_659731.1 | ||||
XM_005256723.2 | Intron | XP_005256780.1 | ||||
XM_011523958.2 | Intron | XP_011522260.1 | ||||
XM_011523959.1 | Intron | XP_011522261.1 | ||||
XM_017024857.1 | Intron | XP_016880346.1 | ||||
XM_017024858.1 | Intron | XP_016880347.1 | ||||
XM_017024859.1 | Intron | XP_016880348.1 |
Set Membership: |
HapMap |