Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 181031 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SAG - S-antigen visual arrestin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000541.4 | Intron | NP_000532.2 | ||||
XM_011511593.2 | Intron | XP_011509895.1 | ||||
XM_011511594.1 | Intron | XP_011509896.1 | ||||
XM_011511596.1 | Intron | XP_011509898.1 | ||||
XM_017004641.1 | Intron | XP_016860130.1 | ||||
XM_017004642.1 | Intron | XP_016860131.1 | ||||
XM_017004643.1 | Intron | XP_016860132.1 |
Set Membership: |
HapMap |