Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607390 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SSBP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SSBP3 - single stranded DNA binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009955.3 | Intron | NP_001009955.1 | ||||
NM_018070.4 | Intron | NP_060540.2 | ||||
NM_145716.3 | Intron | NP_663768.1 | ||||
XM_006710545.3 | Intron | XP_006710608.1 | ||||
XM_017000897.1 | Intron | XP_016856386.1 | ||||
XM_017000898.1 | Intron | XP_016856387.1 | ||||
XM_017000899.1 | Intron | XP_016856388.1 |
SSBP3-AS1 - SSBP3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP |