Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGGTGCAGGAGACCCCGGGCCA[C/T]GCATCCTTCCGGGCAGGAAAATGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609144 MIM: 610864 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLVCR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FLVCR1 - feline leukemia virus subgroup C cellular receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014053.3 | Intron | NP_054772.1 | ||||
XM_011509446.2 | Intron | XP_011507748.1 | ||||
XM_011509447.2 | Intron | XP_011507749.1 | ||||
XM_011509448.1 | Intron | XP_011507750.1 |
FLVCR1-AS1 - FLVCR1 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP |