Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611518 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOCK10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DOCK10 - dedicator of cytokinesis 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290263.1 | Intron | NP_001277192.1 | ||||
NM_014689.2 | Intron | NP_055504.2 | ||||
XM_005246673.4 | Intron | XP_005246730.1 | ||||
XM_006712617.3 | Intron | XP_006712680.1 | ||||
XM_017004441.1 | Intron | XP_016859930.1 | ||||
XM_017004442.1 | Intron | XP_016859931.1 | ||||
XM_017004443.1 | Intron | XP_016859932.1 | ||||
XM_017004444.1 | Intron | XP_016859933.1 | ||||
XM_017004445.1 | Intron | XP_016859934.1 | ||||
XM_017004446.1 | Intron | XP_016859935.1 | ||||
XM_017004447.1 | Intron | XP_016859936.1 |
Set Membership: |
HapMap |