Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612054 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNOT9 - CCR4-NOT transcription complex subunit 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271634.1 | Intron | NP_001258563.1 | ||||
NM_001271635.1 | Intron | NP_001258564.1 | ||||
NM_005444.2 | Intron | NP_005435.1 | ||||
XM_011512138.2 | Intron | XP_011510440.1 | ||||
XM_017005248.1 | Intron | XP_016860737.1 | ||||
XM_017005249.1 | Intron | XP_016860738.1 |