Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616704 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMMD10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
COMMD10 - COMM domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308080.1 | 340 | Intron | NP_001295009.1 | |||
NM_016144.3 | 340 | Intron | NP_057228.1 | |||
XM_011543458.2 | 340 | UTR 5 | XP_011541760.1 | |||
XM_011543460.2 | 340 | UTR 5 | XP_011541762.1 | |||
XM_017009553.1 | 340 | Intron | XP_016865042.1 | |||
XM_017009555.1 | 340 | UTR 5 | XP_016865044.1 | |||
XM_017009556.1 | 340 | UTR 5 | XP_016865045.1 | |||
XM_017009557.1 | 340 | Intron | XP_016865046.1 |