Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607100 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPHP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NPHP1 - nephrocystin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000272.3 | Intron | NP_000263.2 | ||||
NM_001128178.1 | Intron | NP_001121650.1 | ||||
NM_001128179.1 | Intron | NP_001121651.1 | ||||
NM_207181.2 | Intron | NP_997064.2 | ||||
XM_005263676.1 | Intron | XP_005263733.1 | ||||
XM_005263677.1 | Intron | XP_005263734.1 | ||||
XM_005263678.2 | Intron | XP_005263735.1 | ||||
XM_005263679.1 | Intron | XP_005263736.1 | ||||
XM_006712551.1 | Intron | XP_006712614.1 | ||||
XM_006712552.2 | Intron | XP_006712615.1 | ||||
XM_011511244.1 | Intron | XP_011509546.1 | ||||
XM_017004218.1 | Intron | XP_016859707.1 |
Set Membership: |
HapMap |