Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNIH4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CNIH4 - cornichon family AMPA receptor auxiliary protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4742 - microRNA 4742 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR26 - WD repeat domain 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001115113.2 | 1696 | UTR 3 | NP_001108585.2 | |||
NM_025160.6 | 1696 | UTR 3 | NP_079436.4 | |||
XM_005273273.4 | 1696 | UTR 3 | XP_005273330.1 | |||
XM_006711817.3 | 1696 | UTR 3 | XP_006711880.1 | |||
XM_011544281.2 | 1696 | UTR 3 | XP_011542583.1 | |||
XM_011544282.2 | 1696 | UTR 3 | XP_011542584.1 | |||
XM_017002396.1 | 1696 | UTR 3 | XP_016857885.1 |