Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603050 MIM: 607429 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABI1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABI1 - abl interactor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012750.2 | Intron | NP_001012768.1 | ||||
NM_001012751.2 | Intron | NP_001012769.1 | ||||
NM_001012752.2 | Intron | NP_001012770.1 | ||||
NM_001178116.1 | Intron | NP_001171587.1 | ||||
NM_001178119.1 | Intron | NP_001171590.1 | ||||
NM_001178120.1 | Intron | NP_001171591.1 | ||||
NM_001178121.1 | Intron | NP_001171592.1 | ||||
NM_001178122.1 | Intron | NP_001171593.1 | ||||
NM_001178123.1 | Intron | NP_001171594.1 | ||||
NM_001178124.1 | Intron | NP_001171595.1 | ||||
NM_001178125.1 | Intron | NP_001171596.1 | ||||
NM_005470.3 | Intron | NP_005461.2 | ||||
XM_005252332.3 | Intron | XP_005252389.1 | ||||
XM_005252333.3 | Intron | XP_005252390.1 | ||||
XM_006717359.2 | Intron | XP_006717422.1 | ||||
XM_006717360.2 | Intron | XP_006717423.1 | ||||
XM_006717361.2 | Intron | XP_006717424.1 | ||||
XM_011519289.2 | Intron | XP_011517591.1 | ||||
XM_017015458.1 | Intron | XP_016870947.1 | ||||
XM_017015459.1 | Intron | XP_016870948.1 |
PDSS1 - prenyl (decaprenyl) diphosphate synthase, subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |