Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154580 MIM: 608844 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAN2C1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAN2C1 - mannosidase alpha class 2C member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256494.1 | 2999 | Missense Mutation | ATC,GTC | I,V 977 | NP_001243423.1 | |
NM_001256495.1 | 2999 | Missense Mutation | ATC,GTC | I,V 937 | NP_001243424.1 | |
NM_001256496.1 | 2999 | Missense Mutation | ATC,GTC | I,V 861 | NP_001243425.1 | |
NM_006715.3 | 2999 | Missense Mutation | ATC,GTC | I,V 960 | NP_006706.2 | |
XM_005254384.2 | 2999 | Missense Mutation | ATC,GTC | I,V 968 | XP_005254441.1 | |
XM_017022186.1 | 2999 | Missense Mutation | ATC,GTC | I,V 983 | XP_016877675.1 | |
XM_017022187.1 | 2999 | Missense Mutation | ATC,GTC | I,V 952 | XP_016877676.1 | |
XM_017022188.1 | 2999 | Missense Mutation | ATC,GTC | I,V 838 | XP_016877677.1 |
MIR631 - microRNA 631 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEIL1 - nei like DNA glycosylase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |