Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611347 MIM: 604193 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
INTS3 - integrator complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324475.1 | Intron | NP_001311404.1 | ||||
NM_023015.4 | Intron | NP_075391.3 | ||||
XM_005245459.2 | Intron | XP_005245516.1 | ||||
XM_005245461.2 | Intron | XP_005245518.1 | ||||
XM_006711490.3 | Intron | XP_006711553.1 | ||||
XM_011509906.2 | Intron | XP_011508208.1 | ||||
XM_017002132.1 | Intron | XP_016857621.1 |
SLC27A3 - solute carrier family 27 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317929.1 | Intron | NP_001304858.1 | ||||
NM_024330.1 | Intron | NP_077306.1 |
Set Membership: |
HapMap JSNP |